Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2cd2l
Entrez Gene ID
300666
Gene Name
C2CD2-like
Gene Aliases
Tmem24
Chromosome Location
Chr.8: 48641792 - 48652115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011996.1 NP_001011996.1 10 - 11 1744 127
XM_006242919.2 XP_006242981.1 10 - 11 1795 127
GenBank mRNA BC085925.1 - 10 - 11 1744 127

View Full Product Details