The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012465.1 | NP_001012483.1 | 10 - 11 | 1157 | 127 |
GenBank mRNA | AB188302.1 | - | 10 - 11 | 1157 | 127 |