Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Adcyap1
Entrez Gene ID
24166
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
Pacap
Chromosome Location
Chr.9: 121705897 - 121726455 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245674.2 XP_006245736.1 1 - 2 1265 86

View Full Product Details