Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tab3
Entrez Gene ID
317546
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.X: 54035952 - 54090282 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003752051.3 XP_003752099.1 3 - 4 3775 103

View Full Product Details