Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Itgb1bp2
Entrez Gene ID
317258
Gene Name
integrin beta 1 binding protein 2
Gene Aliases
RGD1565015
Chromosome Location
Chr.X: 71342770 - 71347623 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108245.2 NP_001101715.1 2 - 3 170 126
GenBank mRNA FM116382.1 - 2 - 3 129 126
FM118664.1 - 2 - 3 154 126
FQ215476.1 - 2 - 3 165 126
FQ216044.1 - 2 - 3 169 126
FQ216100.1 - 2 - 3 146 126
FQ224925.1 - 2 - 3 167 126

View Full Product Details