Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxo4
Entrez Gene ID
302415
Gene Name
forkhead box O4
Gene Aliases
RGD1561201
Chromosome Location
Chr.X: 71155601 - 71162158 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106943.1 NP_001100413.1 3 - 4 1652 101

View Full Product Details