The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177935.2 | NP_808796.1 | 9 - 10 | 1386 | 143 |
XM_008773261.1 | XP_008771483.1 | 4 - 5 | 1146 | 143 | |
GenBank mRNA | AY216593.1 | - | 9 - 10 | 1386 | 143 |