Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mpv17l2
Entrez Gene ID
290645
Gene Name
MPV17 mitochondrial membrane protein-like 2
Gene Aliases
RGD1308064
Chromosome Location
Chr.16: 20432734 - 20435125 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106072.2 NP_001099542.1 4 - 5 622 68

View Full Product Details