The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012568.2 | NP_036700.1 | 8 - 9 | 1632 | 72 |
XM_003752039.3 | XP_003752087.1 | 9 - 10 | 1256 | 72 | |
XM_008773152.1 | XP_008771374.1 | 8 - 9 | 1625 | 72 | |
XM_008773153.1 | XP_008771375.1 | 8 - 9 | 1038 | 72 | |
GenBank mRNA | AJ310837.1 | - | 8 - 9 | 1081 | 72 |
X61159.1 | - | 8 - 9 | 1632 | 72 |