Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sftpb
Entrez Gene ID
192155
Gene Name
surfactant protein B
Gene Aliases
Sp-b
Chromosome Location
Chr.4: 100166860 - 100175942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138842.1 NP_620197.1 5 - 6 556 134
GenBank mRNA BC072466.1 - 5 - 6 555 134
X14778.1 - 5 - 6 556 134

View Full Product Details