Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Paip2b
Entrez Gene ID
312490
Gene Name
poly(A) binding protein interacting protein 2B
Gene Aliases
RGD1307930
Chromosome Location
Chr.4: 115484676 - 115516303 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_232121.6 XP_232121.4 1 - 2 105 81

View Full Product Details