Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp11a1
Entrez Gene ID
29680
Gene Name
cytochrome P450, family 11, subfamily a, polypeptide 1
Gene Aliases
Cyp11a, Cypxia1, P450scc
Chromosome Location
Chr.8: 62798342 - 62809674 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC089100.1 - 7 - 8 1251 76
M22615.1 - 2 - 3 197 76

View Full Product Details