Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cspg4
Entrez Gene ID
81651
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
Ng2
Chromosome Location
Chr.8: 61532465 - 61567510 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031022.1 NP_112284.1 5 - 6 4531 88
GenBank mRNA X56541.2 - 5 - 6 4531 88

View Full Product Details