Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx21
Entrez Gene ID
100362940
Gene Name
sorting nexin family member 21
Gene Aliases
-
Chromosome Location
Chr.3: 161252636 - 161261880 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762558.1 XP_008760780.1 2 - 3 639 55

View Full Product Details