Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glis2
Entrez Gene ID
302946
Gene Name
GLIS family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.10: 11154460 - 11174905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106978.1 NP_001100448.1 4 - 5 663 69
XM_006245801.2 XP_006245863.1 4 - 5 707 69

View Full Product Details