The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003750771.3 | XP_003750819.1 | 4 - 5 | 1581 | 160 |
XM_008767510.1 | XP_008765732.1 | 3 - 4 | 287 | 160 |