Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srl
Entrez Gene ID
302948
Gene Name
sarcalumenin
Gene Aliases
-
Chromosome Location
Chr.10: 11240141 - 11284325 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750771.3 XP_003750819.1 5 - 6 1695 85
XM_008767510.1 XP_008765732.1 4 - 5 401 85

View Full Product Details