Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snrnp27
Entrez Gene ID
362392
Gene Name
small nuclear ribonucleoprotein 27 (U4/U6.U5)
Gene Aliases
RGD1310925, Ry1
Chromosome Location
Chr.4: 118478075 - 118487633 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108636.1 NP_001102106.1 1 - 2 75 89
GenBank mRNA FQ224589.1 - - 77 89

View Full Product Details