Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp26b1
Entrez Gene ID
312495
Gene Name
cytochrome P450, family 26, subfamily b, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.4: 116261796 - 116278615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181087.2 NP_851601.1 5 - 6 1300 135
GenBank mRNA AY245532.2 - 5 - 6 1300 135

View Full Product Details