The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012737.1 | NP_001012755.1 | 8 - 9 | 1497 | 104 |
GenBank mRNA | AB121031.1 | - | 8 - 9 | 1497 | 104 |