The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008772679.1 | XP_008770901.1 | 6 - 7 | 1822 | 78 |
GenBank mRNA | AY228140.1 | - | 4 - 5 | 607 | 78 |
BC081694.1 | - | 4 - 5 | 851 | 78 |