Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb18
Entrez Gene ID
316614
Gene Name
ankyrin repeat and SOCS box-containing 18
Gene Aliases
-
Chromosome Location
Chr.9: 97085234 - 97151832 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108231.1 NP_001101701.1 1 - 2 201 68

View Full Product Details