The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008381.1 | NP_001008382.1 | 7 - 8 | 453 | 95 |
XM_006245422.2 | XP_006245484.1 | 8 - 9 | 601 | 95 | |
XM_006245423.2 | XP_006245485.1 | 8 - 9 | 800 | 95 | |
XM_006245425.2 | XP_006245487.1 | 8 - 9 | 598 | 95 | |
XM_006245427.2 | XP_006245489.1 | 8 - 9 | 645 | 95 | |
XM_008767274.1 | XP_008765496.1 | 7 - 8 | 571 | 95 | |
XM_008767275.1 | XP_008765497.1 | 7 - 8 | 533 | 95 | |
GenBank mRNA | BC086355.1 | - | 7 - 8 | 453 | 95 |