The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106386.2 | NP_001099856.2 | 4 - 5 | 419 | 103 |
GenBank mRNA | BC158870.1 | - | 4 - 5 | 419 | 103 |