Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fance
Entrez Gene ID
309643
Gene Name
Fanconi anemia, complementation group E
Gene Aliases
RGD1561045
Chromosome Location
Chr.20: 7888927 - 7906142 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191718.1 NP_001178647.1 9 - 10 1548 63

View Full Product Details