Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ggnbp1
Entrez Gene ID
494520
Gene Name
gametogenetin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 5618974 - 5625320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009972.1 NP_001009972.1 1 - 2 548 159
GenBank mRNA AY529104.1 - 1 - 2 548 159
BC105838.1 - 1 - 2 556 159

View Full Product Details