Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggnbp1
Entrez Gene ID
494520
Gene Name
gametogenetin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 5618974 - 5625320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009972.1 NP_001009972.1 3 - 4 764 89
XM_006256125.2 XP_006256187.1 3 - 4 822 89
GenBank mRNA AY529104.1 - 3 - 4 764 89
BC105838.1 - 3 - 4 772 89

View Full Product Details