Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnah8
Entrez Gene ID
207117
Gene Name
dynein, axonemal, heavy chain 8
Gene Aliases
Dlp8, Dnahc8
Chromosome Location
Chr.20: 9301314 - 9560805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772852.1 XP_008771074.1 45 - 46 6694 137
GenBank mRNA D26499.1 - 3 - 4 379 137

View Full Product Details