The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037214.1 | NP_001032291.1 | 2 - 2 | 1190 | 134 |
XM_008764947.1 | XP_008763169.1 | 3 - 3 | 1313 | 134 | |
XM_008764948.1 | XP_008763170.1 | 2 - 2 | 1054 | 134 | |
GenBank mRNA | BC107668.1 | - | 2 - 2 | 1190 | 134 |