Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klc1
Entrez Gene ID
171041
Gene Name
kinesin light chain 1
Gene Aliases
Kns2
Chromosome Location
Chr.6: 136329840 - 136373716 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081972.1 NP_001075441.1 4 - 5 688 109
NM_001081973.1 NP_001075442.1 4 - 5 711 109
NM_001081974.1 NP_001075443.1 4 - 5 697 109
XM_006240621.2 XP_006240683.1 4 - 5 1281 109
XM_006240622.2 XP_006240684.1 4 - 5 1296 109
XM_006240623.2 XP_006240685.1 4 - 5 1282 109
XM_006240625.2 XP_006240687.1 4 - 5 1177 109
XM_006240626.2 XP_006240688.1 4 - 5 1176 109
XM_006240627.2 XP_006240689.1 4 - 5 1177 109
XM_006240628.2 XP_006240690.1 4 - 5 1177 109
XM_006240629.2 XP_006240691.1 4 - 5 1176 109
XM_006240630.2 XP_006240692.1 4 - 5 1177 109
GenBank mRNA M75146.1 - 4 - 5 688 109
M75147.1 - 4 - 5 711 109
M75148.1 - 4 - 5 697 109

View Full Product Details