Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgs17
Entrez Gene ID
308118
Gene Name
regulator of G-protein signaling 17
Gene Aliases
-
Chromosome Location
Chr.1: 42491566 - 42587734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107459.1 NP_001100929.1 4 - 5 462 128
XM_006227854.2 XP_006227916.1 3 - 4 403 128
XM_006227855.2 XP_006227917.1 3 - 4 424 128

View Full Product Details