Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem184c
Entrez Gene ID
291946
Gene Name
transmembrane protein 184C
Gene Aliases
Tmem34
Chromosome Location
Chr.19: 34041165 - 34057165 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178330.1 NP_847900.1 9 - 10 1507 84
XM_006255422.2 XP_006255484.1 10 - 11 1219 84
XM_008772475.1 XP_008770697.1 10 - 11 2118 84
GenBank mRNA AY228474.1 - 9 - 10 1507 84
BC089112.1 - 9 - 10 1517 84

View Full Product Details