The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009639.1 | NP_001009639.1 | 3 - 4 | 445 | 62 |
XM_006255464.2 | XP_006255526.1 | 3 - 4 | 475 | 62 | |
GenBank mRNA | BC087585.1 | - | 3 - 4 | 445 | 62 |
FQ213895.1 | - | 2 - 3 | 587 | 62 | |
FQ214516.1 | - | 3 - 4 | 459 | 62 | |
FQ214740.1 | - | 3 - 4 | 449 | 62 | |
FQ216797.1 | - | 3 - 4 | 470 | 62 | |
FQ217207.1 | - | 3 - 4 | 446 | 62 |