Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lphn1
Entrez Gene ID
65096
Gene Name
latrophilin 1
Gene Aliases
CL1BA
Chromosome Location
Chr.19: 25029037 - 25069146 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022962.1 NP_075251.1 19 - 20 3590 93
GenBank mRNA AF081144.1 - 18 - 19 3575 93
AF081145.1 - 18 - 19 3575 93
AF081146.1 - 19 - 20 3590 93
AF081147.1 - 19 - 20 3590 93
AF111099.1 - 20 - 21 3916 93
U72487.1 - 18 - 19 3392 93
U78105.1 - 18 - 19 3717 93

View Full Product Details