Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Calb2
Entrez Gene ID
117059
Gene Name
calbindin 2
Gene Aliases
-
Chromosome Location
Chr.19: 41475795 - 41509630 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053988.1 NP_446440.1 6 - 7 533 70
XM_008772466.1 XP_008770688.1 7 - 8 712 70
GenBank mRNA BC087603.1 - 6 - 7 503 70
X66974.1 - 6 - 7 533 70

View Full Product Details