Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Afg3l1
Entrez Gene ID
361436
Gene Name
AFG3(ATPase family gene 3)-like 1 (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.19: 56272171 - 56299323 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108456.3 NP_001101926.3 10 - 11 1321 83
GenBank mRNA BC168848.1 - 10 - 11 1214 83

View Full Product Details