Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc29
Entrez Gene ID
291944
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.19: 33206531 - 33424955 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013890.1 NP_001013912.1 6 - 7 757 62
GenBank mRNA BC078968.1 - 6 - 7 757 62

View Full Product Details