Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gas8
Entrez Gene ID
361438
Gene Name
growth arrest specific 8
Gene Aliases
-
Chromosome Location
Chr.19: 56315862 - 56335873 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039030.1 NP_001034119.1 10 - 11 1310 122
XM_006255777.2 XP_006255839.1 10 - 11 1351 122
XM_008772635.1 XP_008770857.1 7 - 8 942 122
XM_008772636.1 XP_008770858.1 7 - 8 878 122
XM_008772637.1 XP_008770859.1 7 - 8 870 122
GenBank mRNA BC099759.1 - 10 - 11 1310 122

View Full Product Details