Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gas8
Entrez Gene ID
361438
Gene Name
growth arrest specific 8
Gene Aliases
-
Chromosome Location
Chr.19: 56315862 - 56335873 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039030.1 NP_001034119.1 3 - 4 305 95
XM_006255777.2 XP_006255839.1 3 - 4 346 95
XM_006255779.2 XP_006255841.1 3 - 4 887 95
GenBank mRNA BC099759.1 - 3 - 4 305 95

View Full Product Details