Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Terf2
Entrez Gene ID
361403
Gene Name
telomeric repeat binding factor 2
Gene Aliases
-
Chromosome Location
Chr.19: 39271992 - 39301500 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108448.1 NP_001101918.1 3 - 4 511 154
NM_001242355.1 NP_001229284.1 3 - 4 474 154
XM_006255564.2 XP_006255626.1 3 - 4 1118 154
GenBank mRNA BC160846.1 - 3 - 4 629 154
FQ212165.1 - 3 - 4 526 154
HQ651915.1 - 3 - 4 474 154

View Full Product Details