The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108448.1 | NP_001101918.1 | 6 - 7 | 856 | 79 |
NM_001242355.1 | NP_001229284.1 | 6 - 7 | 819 | 79 | |
XM_006255564.2 | XP_006255626.1 | 6 - 7 | 1463 | 79 | |
GenBank mRNA | BC160846.1 | - | 6 - 7 | 974 | 79 |
HQ651915.1 | - | 6 - 7 | 819 | 79 |