Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfpm1
Entrez Gene ID
691504
Gene Name
zinc finger protein, multitype 1
Gene Aliases
-
Chromosome Location
Chr.19: 55094585 - 55150994 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242627.1 NP_001229556.1 5 - 6 580 112
XM_008772646.1 XP_008770868.1 4 - 5 4048 112
XM_008772647.1 XP_008770869.1 4 - 5 4048 112
XM_008772649.1 XP_008770871.1 3 - 4 522 112

View Full Product Details