The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242627.1 | NP_001229556.1 | 9 - 10 | 1257 | 66 |
XM_008772646.1 | XP_008770868.1 | 8 - 9 | 4737 | 66 | |
XM_008772647.1 | XP_008770869.1 | 8 - 9 | 4725 | 66 | |
XM_008772649.1 | XP_008770871.1 | 7 - 8 | 1211 | 66 |