The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083313.1 | NP_001076782.1 | 5 - 6 | 451 | 68 |
GenBank mRNA | CK358527.1 | - | 5 - 6 | 430 | 68 |