The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007740.1 | NP_001007741.1 | 8 - 9 | 953 | 88 |
XM_006256460.2 | XP_006256522.1 | 8 - 9 | 1076 | 88 | |
GenBank mRNA | BC079150.1 | - | 8 - 9 | 953 | 88 |
FQ211877.1 | - | 8 - 9 | 966 | 88 |