Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Man1a1
Entrez Gene ID
294410
Gene Name
mannosidase, alpha, class 1A, member 1
Gene Aliases
Aa2-166, Man1a
Chromosome Location
Chr.20: 35257688 - 35450132 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033656.1 NP_001028828.1 9 - 10 1843 137
XM_006256497.1 XP_006256559.1 9 - 10 1715 137

View Full Product Details