Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dtnbp1
Entrez Gene ID
641528
Gene Name
dystrobrevin binding protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 20090136 - 20182332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037664.1 NP_001032753.1 8 - 9 927 101
GenBank mRNA AY623026.1 - - 1501 101
BC088267.1 - 8 - 9 927 101

View Full Product Details