Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmpr
Entrez Gene ID
117533
Gene Name
guanosine monophosphate reductase
Gene Aliases
Gmpr1
Chromosome Location
Chr.17: 19543274 - 19580929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057188.1 NP_476536.1 4 - 5 541 83
GenBank mRNA AF090867.1 - 4 - 5 541 83

View Full Product Details