The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126083.1 | NP_001119555.1 | 2 - 3 | 295 | 77 |
GenBank mRNA | BC162005.1 | - | 2 - 3 | 295 | 77 |