Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cage1
Entrez Gene ID
306872
Gene Name
cancer antigen 1
Gene Aliases
Ctag3
Chromosome Location
Chr.17: 27452506 - 27490853 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012052.1 NP_001012052.1 7 - 8 2314 123
XM_006253808.2 XP_006253870.1 7 - 8 2301 123
XM_006253809.2 XP_006253871.1 6 - 7 1992 123
XM_006253810.2 XP_006253872.1 6 - 7 2184 123
XM_006253811.2 XP_006253873.1 6 - 7 2268 123
GenBank mRNA BC081934.1 - 7 - 8 2314 123
FQ211932.1 - 6 - 7 2272 123

View Full Product Details